Gene Transcripts

Gene Symbol
Parp9
Entrez Gene ID
80285
Gene Name
poly (ADP-ribose) polymerase family, member 9
Gene Aliases
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9
Chromosome Location
Chr. 16: 35938470 - 35972621 on Build GRCm38
UniGene ID
Mm.49074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030253.3 NP_084529.1 5 1781
GenBank mRNA AK037903.1 - 5 1348
AK050032.1 - 5 1225
BC003281.1 - 5 1223 3581841
BC010312.1 - 4 1206
BC070466.1 - 5 1781 30615225

View Full Product Details