Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033509.3 | NP_277044.1 | 7 | 1699 | |
GenBank mRNA | AF365875.1 | - | 7 | 1463 | |
AK033128.1 | - | 8 | 1739 | ||
AK046036.1 | - | 7 | 1716 | ||
AK049613.1 | - | 1714 | |||
AK129311.1 | - | 1699 | |||
AY035370.3 | - | 7 | 1687 | ||
BC052195.1 | - | 7 | 1699 | 6509008 | |
BC075636.1 | - | 2556 |