Gene Transcripts

Gene Symbol
Olfr166
Entrez Gene ID
259071
Gene Name
olfactory receptor 166
Gene Aliases
MOR270-1
Chromosome Location
Chr. 16: 19486840 - 19487778 on Build GRCm38
UniGene ID
Mm.377822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147068.1 NP_667279.1 1 300
GenBank mRNA BC116856.1 - 1 347
BC119189.1 - 1 347 8733938

View Full Product Details