Gene Transcripts

Gene Symbol
Ppp1r1a
Entrez Gene ID
58200
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 1A
Gene Aliases
0610038N18Rik, I-1
Chromosome Location
Chr. 15: 103530279 - 103538005 on Build GRCm38
UniGene ID
Mm.143788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021391.3 NP_067366.1 2, 3 267
GenBank mRNA AF281676.1 - 2, 3 157
BC025123.1 - 2, 3 236 4019442
CA321941.1 - 2, 3 267

View Full Product Details