Gene Transcripts

Gene Symbol
Fam131a
Entrez Gene ID
78408
Gene Name
family with sequence similarity 131, member A
Gene Aliases
2900046G09Rik
Chromosome Location
Chr. 16: 20693096 - 20703036 on Build GRCm38
UniGene ID
Mm.196512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133778.2 NP_598539.1 2, 3 270
GenBank mRNA AK049983.1 - 2, 3 271
BC092532.1 - 2, 3 301 6594874

View Full Product Details