Gene Transcripts

Gene Symbol
Olfr686
Entrez Gene ID
259072
Gene Name
olfactory receptor 686
Gene Aliases
MOR35-1
Chromosome Location
Chr. 7: 105203388 - 105204341 on Build GRCm38
UniGene ID
Mm.377823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147069.1 NP_667280.1 1 144
GenBank mRNA BC132004.1 - 1 179
BC132006.1 - 1 179 40130283

View Full Product Details