Gene Transcripts

Gene Symbol
TMPRSS13
Entrez Gene ID
84000
Gene Name
transmembrane protease, serine 13
Gene Aliases
MSP, MSPL, MSPS, TMPRSS11
Chromosome Location
Chr. 11: 117900641 - 117929453 on Build GRCh38
UniGene ID
Hs.266308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077263.2 NP_001070731.1 13 2442
NM_001206789.1 NP_001193718.1 12 2337
GenBank mRNA AB048797.1 - 12 2289
BC018715.1 - 14 2179
BC114928.1 - 13 2361 40028749

View Full Product Details