Gene Transcripts

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
Chromosome Location
Chr. 2: 190343470 - 190371665 on Build GRCh38
UniGene ID
Hs.32309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128928.1 NP_001122400.1 7 1515
NM_002194.3 NP_002185.1 6 1371
XM_005246532.1 XP_005246589.1 6 1152
GenBank mRNA AB451298.1 - 5 815
AK093560.1 - 8 1473
BC015496.1 - 5 881 3845956
BC106006.1 - 6 915 4472604
CR456840.1 - 5 815
CR541707.1 - 5 815
KJ535023.1 - 5 813
KJ535042.1 - 5 814
KU178146.1 - 5 815
KU178147.1 - 4 564
L08488.1 - 6 1141

View Full Product Details