Gene Transcripts
- Gene Symbol
- PAXIP1
- Entrez Gene ID
- 22976
- Gene Name
- PAX interacting protein 1
- Gene Aliases
- CAGF28, CAGF29, PACIP1, PAXIP1L, PTIP, TNRC2
- Chromosome Location
- Chr. 7: 154943690 - 155003393 on Build GRCh38
- UniGene ID
- Hs.443881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007349.3 |
NP_031375.3
|
19, 20 |
3178 |
|
XM_005249539.1 |
XP_005249596.1
|
20, 21 |
3317 |
|
XM_011515982.2 |
XP_011514284.1
|
19, 20 |
3292 |
|
XM_011515983.1 |
XP_011514285.1
|
19, 20 |
3178 |
|
XM_017011881.1 |
XP_016867370.1
|
20, 21 |
4490 |
|
GenBank mRNA |
AK093584.1 |
- |
13, 14 |
1871 |
|
AK123600.1 |
- |
20, 21 |
3317 |
|
AK126674.1 |
- |
11, 12 |
3257 |
|
AK293744.1 |
- |
17, 18 |
3010 |
|
BC008328.1 |
- |
10, 11 |
1102 |
|
BC033781.1 |
- |
14, 15 |
2419 |
4509264 |
BX538201.1 |
- |
13, 14 |
1954 |
|
BX640616.1 |
- |
19, 20 |
5076 |
|
U80735.1 |
- |
15, 16 |
2161 |
|
View Full Product Details