Gene Transcripts
- Gene Symbol
- GPR107
- Entrez Gene ID
- 57720
- Gene Name
- G protein-coupled receptor 107
- Gene Aliases
- GCDRP, LUSTR1, bA138E2.2
- Chromosome Location
- Chr. 9: 130053706 - 130140169 on Build GRCh38
- UniGene ID
- Hs.512461
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136557.1 |
NP_001130029.1
|
4, 5 |
622 |
|
NM_001136558.1 |
NP_001130030.1
|
4, 5 |
622 |
|
NM_020960.4 |
NP_066011.2
|
4, 5 |
622 |
|
XM_017014953.1 |
XP_016870442.1
|
4, 5 |
583 |
|
XM_017014954.1 |
XP_016870443.1
|
4, 5 |
583 |
|
GenBank mRNA |
AB046844.1 |
- |
4, 5 |
393 |
|
AF376725.1 |
- |
4, 5 |
403 |
|
AK026244.1 |
- |
4, 5 |
407 |
|
AK225456.1 |
- |
4, 5 |
407 |
|
AK293092.1 |
- |
4, 5 |
403 |
|
AL834359.1 |
- |
3, 4 |
266 |
|
BC110518.2 |
- |
4, 5 |
403 |
40033360 |
BC143655.1 |
- |
4, 5 |
418 |
|
View Full Product Details