Gene Transcripts

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
Chromosome Location
Chr. 4: 76011180 - 76112802 on Build GRCh38
UniGene ID
Hs.24976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130016.2 NP_001123488.1 2, 3 211
NM_001130017.2 NP_001123489.1 2, 3 227
NM_001179.5 NP_001170.2 2, 3 211
XM_005262997.1 XP_005263054.1 1, 2 86
XM_005262999.1 XP_005263056.1 1, 2 86
XM_005263003.1 XP_005263060.1 1, 2 86
XM_005263004.1 XP_005263061.1 1, 2 86
XM_006714220.1 XP_006714283.1 1, 2 86
XM_011531971.1 XP_011530273.1 1, 2 86
XM_017008206.1 XP_016863695.1 1, 2 86
XM_017008207.1 XP_016863696.1 1, 2 86
XM_017008209.1 XP_016863698.1 1, 2 86
GenBank mRNA AK225860.1 - 2, 3 207
AK295305.1 - 2, 3 193
AK308810.1 - 2, 3 193
AY543657.1 - 2, 3 203
AY543658.1 - 3, 4 237
AY543659.1 - 4, 5 318
AY543660.1 - 2, 3 196
BC008397.1 - 2, 3 198 4244549
BC017913.1 - 2, 3 198 4292261
BT007249.1 - 1, 2 77
CR541664.1 - 1, 2 77
DB451386.1 - 3, 4 257
U47054.1 - 2, 3 107

View Full Product Details