Gene Transcripts
- Gene Symbol
- TESK2
- Entrez Gene ID
- 10420
- Gene Name
- testis-specific kinase 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 45343883 - 45491342 on Build GRCh38
- UniGene ID
- Hs.591499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007170.2 |
NP_009101.2
|
2, 3 |
633 |
|
XM_005270355.3 |
XP_005270412.1
|
2, 3 |
605 |
|
XM_006710287.2 |
XP_006710350.1
|
3, 4 |
673 |
|
XM_011540497.1 |
XP_011538799.1
|
4, 5 |
706 |
|
XM_011540500.2 |
XP_011538802.1
|
2, 3 |
605 |
|
XM_017000047.1 |
XP_016855536.1
|
3, 4 |
838 |
|
XM_017000048.1 |
XP_016855537.1
|
2, 3 |
807 |
|
XM_017000049.1 |
XP_016855538.1
|
2, 3 |
807 |
|
GenBank mRNA |
AB057597.1 |
- |
2, 3 |
625 |
|
AJ132545.1 |
- |
2, 3 |
625 |
|
AK027573.1 |
- |
2, 3 |
607 |
|
AK299269.1 |
- |
2, 3 |
628 |
|
AK302494.1 |
- |
2, 3 |
665 |
|
BC022194.1 |
- |
2, 3 |
613 |
|
BC033085.1 |
- |
1, 2 |
244 |
5425494 |
View Full Product Details