Gene Transcripts
- Gene Symbol
- FTCD
- Entrez Gene ID
- 10841
- Gene Name
- formimidoyltransferase cyclodeaminase
- Gene Aliases
- LCHC1
- Chromosome Location
- Chr. 21: 46135981 - 46156481 on Build GRCh38
- UniGene ID
- Hs.415846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320412.1 |
NP_001307341.1
|
8, 9 |
1006 |
|
NM_006657.2 |
NP_006648.1
|
8, 9 |
1006 |
|
NM_206965.1 |
NP_996848.1
|
8, 9 |
1006 |
|
XM_006723961.3 |
XP_006724024.2
|
8, 9 |
1297 |
|
XM_006723962.3 |
XP_006724025.2
|
8, 9 |
1905 |
|
XM_011529434.2 |
XP_011527736.1
|
8, 9 |
1920 |
|
XM_011529435.2 |
XP_011527737.1
|
8, 9 |
1304 |
|
XM_011529436.2 |
XP_011527738.1
|
8, 9 |
1905 |
|
XM_011529437.2 |
XP_011527739.1
|
8, 9 |
1299 |
|
XM_011529439.2 |
XP_011527741.1
|
6, 7 |
709 |
|
XM_011529440.2 |
XP_011527742.1
|
8, 9 |
1291 |
|
GenBank mRNA |
AF169017.1 |
- |
8, 9 |
1006 |
|
AF289021.1 |
- |
8, 9 |
1006 |
|
AF289022.1 |
- |
8, 9 |
1006 |
|
AF289023.1 |
- |
8, 9 |
1006 |
|
AF289024.1 |
- |
8, 9 |
1031 |
|
BC052248.1 |
- |
8, 9 |
1002 |
6270063 |
BC136383.1 |
- |
8, 9 |
998 |
9020370 |
BC136395.1 |
- |
8, 9 |
998 |
9020382 |
View Full Product Details