Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166434.2 | NP_001159906.1 | 2 | 225 | |
NM_002215.3 | NP_002206.2 | 4 | 386 | ||
GenBank mRNA | AK292750.1 | - | 4 | 356 | |
AK298398.1 | - | 4 | 385 | ||
AK298423.1 | - | 4 | 378 | ||
AK298455.1 | - | 2 | 211 | ||
AK314198.1 | - | 4 | 356 | ||
BC069464.1 | - | 4 | 380 | 7262464 | |
BX496392.1 | - | 4 | 387 | ||
DB188130.1 | - | 1 | 500 | ||
X16260.1 | - | 2 | 135 | ||
X63652.1 | - | 4 | 380 |