Gene Transcripts
- Gene Symbol
- SND1
- Entrez Gene ID
- 27044
- Gene Name
- staphylococcal nuclease and tudor domain containing 1
- Gene Aliases
- TDRD11, p100
- Chromosome Location
- Chr. 7: 127652119 - 128092607 on Build GRCh38
- UniGene ID
- Hs.122523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014390.2 |
NP_055205.2
|
|
3503 |
|
GenBank mRNA |
U22055.1 |
- |
|
3469 |
|
View Full Product Details