Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014310.3 | NP_055125.2 | 3 | 753 | |
XM_005261442.4 | XP_005261499.1 | 3 | 613 | ||
XM_011530040.2 | XP_011528342.1 | 4 | 1584 | ||
XM_017028701.1 | XP_016884190.1 | 2 | 3362 | ||
XM_017028702.1 | XP_016884191.1 | 3 | 1671 | ||
GenBank mRNA | AF279143.1 | - | 2 | 584 | |
AL831936.1 | - | 2 | 465 | ||
BC013419.2 | - | 3 | 755 | 3604196 | |
CR456477.1 | - | 3 | 668 |