Gene Transcripts

Gene Symbol
TUBG2
Entrez Gene ID
27175
Gene Name
tubulin gamma 2
Gene Aliases
Chromosome Location
Chr. 17: 42659248 - 42667006 on Build GRCh38
UniGene ID
Hs.279669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320509.1 NP_001307438.1 4 640
NM_016437.2 NP_057521.1 4 640
XM_011524614.2 XP_011522916.1 4 655
XM_017024453.1 XP_016879942.1 4 635
XM_017024454.1 XP_016879943.1 4 577
XM_017024455.1 XP_016879944.1 4 654
XM_017024456.1 XP_016879945.1 4 654
XM_017024457.1 XP_016879946.1 4 441
GenBank mRNA AF225971.1 - 4 633
AK022324.1 - 4 583
AK225134.1 - 4 599
AK310562.1 - 2 157
BC009670.2 - 4 492 3891214
BC051890.1 - 4 583 5726689
BC108739.1 - 4 601 5242733
BI819869.1 - 4 588

View Full Product Details