Gene Transcripts

Gene Symbol
AQP2
Entrez Gene ID
359
Gene Name
aquaporin 2
Gene Aliases
AQP-CD, WCH-CD
Chromosome Location
Chr. 12: 49950741 - 49958881 on Build GRCh38
UniGene ID
Hs.130730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000486.5 NP_000477.1 4 1632
GenBank mRNA BC042496.1 - 4 1627

View Full Product Details