Gene Transcripts
- Gene Symbol
- ULK3
- Entrez Gene ID
- 25989
- Gene Name
- unc-51 like kinase 3
- Gene Aliases
- Chromosome Location
- Chr. 15: 74836116 - 74843211 on Build GRCh38
- UniGene ID
- Hs.513034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099436.3 |
NP_001092906.3
|
14, 15 |
1443 |
|
NM_001284365.2 |
NP_001271294.1
|
14, 15 |
1366 |
|
NR_104307.2 |
|
13, 14 |
1624 |
|
NR_104309.2 |
|
13, 14 |
1605 |
|
XM_005254289.2 |
XP_005254346.1
|
15, 16 |
1458 |
|
XM_017022069.1 |
XP_016877558.1
|
15, 16 |
1496 |
|
XM_017022070.1 |
XP_016877559.1
|
15, 16 |
1509 |
|
GenBank mRNA |
AK075047.1 |
- |
12, 13 |
1454 |
|
AK293181.1 |
- |
14, 15 |
1296 |
|
AK294245.1 |
- |
14, 15 |
1373 |
|
AK299380.1 |
- |
14, 15 |
1370 |
|
AL117482.1 |
- |
13, 14 |
1759 |
|
AL360256.1 |
- |
9, 10 |
1036 |
|
BC036117.1 |
- |
13, 14 |
1595 |
|
BC056423.1 |
- |
13, 14 |
1523 |
|
View Full Product Details