Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019071.2 | NP_061944.2 | 3 | 320 | |
NM_198267.1 | NP_938008.1 | 3 | 320 | ||
XM_017012369.1 | XP_016867858.1 | 3 | 328 | ||
GenBank mRNA | AF074968.1 | - | 3 | 297 | |
AF161419.1 | - | 3 | 310 | ||
AF180298.1 | - | 3 | 287 | ||
AK000096.1 | - | 3 | 321 | ||
AK291905.1 | - | 3 | 298 | ||
AK293682.1 | - | 2 | 206 | ||
AL603623.1 | - | 3 | 297 | ||
AY007790.1 | - | 3 | 301 | ||
BC009776.1 | - | 3 | 275 | ||
BC009777.1 | - | 3 | 308 | ||
BC010851.1 | - | 3 | 267 | ||
BC062634.1 | - | 3 | 283 | ||
BC073865.1 | - | 3 | 288 | ||
BC093091.1 | - | 3 | 304 | ||
BC093689.1 | - | 3 | 280 | ||
BC101609.1 | - | 3 | 280 | ||
BC143316.1 | - | 3 | 280 |