Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032021.2 | NP_114410.1 | 1 | 778 | |
GenBank mRNA | AF247167.1 | - | 1 | 777 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011542615.2 | XP_011540917.1 | 24 | 9172 | |
XM_011542616.2 | XP_011540918.1 | 24 | 6941 | ||
XM_011542617.2 | XP_011540919.1 | 25 | 6827 | ||
XM_017017237.1 | XP_016872726.1 | 24 | 7130 | ||
XM_017017238.1 | XP_016872727.1 | 25 | 9313 |