Gene Transcripts
- Gene Symbol
- CSMD2
- Entrez Gene ID
- 114784
- Gene Name
- CUB and Sushi multiple domains 2
- Gene Aliases
- dJ1007G16.1, dJ1007G16.2, dJ947L8.1
- Chromosome Location
- Chr. 1: 33513999 - 34165842 on Build GRCh38
- UniGene ID
- Hs.656915
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281956.1 |
NP_001268885.1
|
12, 13 |
1835 |
|
NM_052896.4 |
NP_443128.2
|
12, 13 |
1567 |
|
XM_017000185.1 |
XP_016855674.1
|
12, 13 |
1829 |
|
XM_017000186.1 |
XP_016855675.1
|
12, 13 |
1829 |
|
XM_017000187.1 |
XP_016855676.1
|
12, 13 |
1658 |
|
XM_017000188.1 |
XP_016855677.1
|
12, 13 |
1567 |
|
XM_017000189.1 |
XP_016855678.1
|
12, 13 |
1829 |
|
XM_017000191.1 |
XP_016855680.1
|
12, 13 |
1829 |
|
XM_017000192.1 |
XP_016855681.1
|
12, 13 |
1829 |
|
XM_017000193.1 |
XP_016855682.1
|
12, 13 |
1829 |
|
XM_017000194.1 |
XP_016855683.1
|
12, 13 |
1567 |
|
GenBank mRNA |
AB212622.1 |
- |
12, 13 |
1835 |
|
AY210418.1 |
- |
12, 13 |
1567 |
|
BC031871.2 |
- |
6, 7 |
895 |
|
View Full Product Details