Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145046.4 | NP_659483.2 | 9 | 1184 | |
GenBank mRNA | AB576176.1 | - | 9 | 1121 | |
AK058084.1 | - | 9 | 1176 | ||
BC014595.2 | - | 9 | 1184 | 4822010 | |
DB510196.1 | - | 2 | 224 |