Gene Transcripts
- Gene Symbol
- EPB41L4A
- Entrez Gene ID
- 64097
- Gene Name
- erythrocyte membrane protein band 4.1 like 4A
- Gene Aliases
- EPB41L4, NBL4
- Chromosome Location
- Chr. 5: 112141829 - 112419490 on Build GRCh38
- UniGene ID
- Hs.584954
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022140.3 |
NP_071423.3
|
1, 2 |
372 |
|
XM_011543530.1 |
XP_011541832.1
|
1, 2 |
549 |
|
XM_011543531.2 |
XP_011541833.1
|
1, 2 |
549 |
|
XM_011543532.1 |
XP_011541834.1
|
1, 2 |
549 |
|
XM_011543533.1 |
XP_011541835.1
|
1, 2 |
549 |
|
XM_017009689.1 |
XP_016865178.1
|
1, 2 |
549 |
|
GenBank mRNA |
AB030240.1 |
- |
1, 2 |
101 |
|
BC031042.1 |
- |
1, 2 |
372 |
|
BC114632.1 |
- |
1, 2 |
198 |
|
BC114942.1 |
- |
1, 2 |
193 |
|
View Full Product Details