Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033647.4 | NP_387467.2 | 4 | 1697 | |
NR_135080.1 | 4 | 1697 | |||
NR_135081.1 | 4 | 1697 | |||
XM_005269234.2 | XP_005269291.1 | 4 | 1675 | ||
XM_017020230.1 | XP_016875719.1 | 4 | 1675 | ||
GenBank mRNA | AF319995.1 | - | 4 | 1638 | |
AK290894.1 | - | 4 | 1662 | ||
BC143409.1 | - | 4 | 1663 | ||
BC143411.1 | - | 4 | 1663 |