Gene Transcripts

Gene Symbol
SLC46A2
Entrez Gene ID
57864
Gene Name
solute carrier family 46 member 2
Gene Aliases
Ly110, TSCOT
Chromosome Location
Chr. 9: 112878920 - 112890981 on Build GRCh38
UniGene ID
Hs.512668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033051.3 NP_149040.3 4 1925
GenBank mRNA AF242557.1 - 4 1925
AK055571.1 - 5 1865
BC048285.1 - 4 1777 5213610

View Full Product Details