Gene Transcripts

Gene Symbol
RNF19A
Entrez Gene ID
25897
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
RNF19
Chromosome Location
Chr. 8: 100257059 - 100336218 on Build GRCh38
UniGene ID
Hs.292882
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280539.1 NP_001267468.1 11 3782
NM_015435.4 NP_056250.3 11 3813
NM_183419.3 NP_904355.1 10 3889
XM_005250853.3 XP_005250910.1 10 3851
XM_011516956.2 XP_011515258.1 9 3712
XM_017013302.1 XP_016868791.1 11 3974
XM_017013303.1 XP_016868792.1 11 3884
XM_017013304.1 XP_016868793.1 9 3669
XM_017013305.1 XP_016868794.1 8 3369
GenBank mRNA AB029316.1 - 11 3813
AK023455.1 - 1 1489
AK027070.1 - 5 2232
AL110253.1 - 1 1287
AL122096.1 - 3 1929
BC043648.1 - 1 1474

View Full Product Details