Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005505.2 | NP_001005505.1 | 3 | 485 | |
NM_001174051.2 | NP_001167522.1 | 3 | 485 | ||
NM_001291101.1 | NP_001278030.1 | 3 | 281 | ||
NM_006030.3 | NP_006021.2 | 3 | 485 | ||
XM_005265581.4 | XP_005265638.1 | 3 | 359 | ||
XM_011534243.2 | XP_011532545.1 | 3 | 359 | ||
GenBank mRNA | AB011130.1 | - | 3 | 312 | |
AF040709.1 | - | 3 | 491 | ||
AF042792.1 | - | 4 | 472 | ||
AF042793.1 | - | 3 | 288 | ||
AJ251367.1 | - | 3 | 311 | ||
AJ251368.1 | - | 3 | 311 | ||
AK310138.1 | - | 4 | 475 | ||
BC152438.1 | - | 3 | 312 | 8862560 |