Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004628.4 | NP_004619.3 | 9 | 1683 | |
NR_027299.1 | 8 | 1559 | |||
XM_011534093.2 | XP_011532395.1 | 9 | 1640 | ||
GenBank mRNA | AK222844.1 | - | 8 | 1376 | |
AK289761.1 | - | 9 | 1612 | ||
AK295711.1 | - | 9 | 1501 | ||
AK311039.1 | - | 8 | 1489 | ||
BC016620.1 | - | 9 | 1594 | 4509957 | |
D21089.1 | - | 9 | 1684 | ||
EU530523.1 | - | 1 | 136 | ||
EU530524.1 | - | 1 | 24 | ||
EU530525.1 | - | 1 | 68 | ||
EU530526.1 | - | 1 | 24 | ||
EU530527.1 | - | 1 | 68 | ||
EU530529.1 | - | 1 | 136 | ||
EU530532.1 | - | 1 | 41 | ||
EU530534.1 | - | 1 | 136 | ||
HM113488.1 | - | 9 | 1583 | ||
KJ534962.1 | - | 9 | 1579 | ||
KJ535085.1 | - | 8 | 1456 | ||
X65024.1 | - | 8 | 1419 |