Gene Transcripts

Gene Symbol
SULT1C2
Entrez Gene ID
6819
Gene Name
sulfotransferase family 1C member 2
Gene Aliases
ST1C1, ST1C2, SULT1C1, humSULTC2
Chromosome Location
Chr. 2: 108288639 - 108309915 on Build GRCh38
UniGene ID
Hs.436123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001056.3 NP_001047.1 8 2072
NM_176825.2 NP_789795.1 9 2105
XM_005264013.3 XP_005264070.1 8 1893
GenBank mRNA AB008164.1 - 8 1742
BM849361.1 - 1 395
R86241.1 - 1 220

View Full Product Details