Gene Transcripts

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
Chromosome Location
Chr. 17: 76736565 - 76803805 on Build GRCh38
UniGene ID
Hs.732019
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242532.1 NP_001229461.1 13 2058
NM_001242533.1 NP_001229462.1 14 1942
NM_001242534.1 NP_001229463.1 14 1804
NM_001242535.1 NP_001229464.1 14 1753
NM_001242536.1 NP_001229465.1 12 1786
NM_001242537.1 NP_001229466.1 12 1597
NM_024311.3 NP_077287.1 14 1660
XM_017025066.1 XP_016880555.1 14 1335
XM_017025067.1 XP_016880556.1 14 1393
XM_017025068.1 XP_016880557.1 14 1805
XM_017025070.1 XP_016880559.1 12 1493
GenBank mRNA AF015184.1 - 14 1372
AF015185.1 - 12 1403
AF015186.1 - 14 1516
AK000233.1 - 13 1440
AK025849.1 - 13 1195
AK308058.1 - 12 1241
BC002753.2 - 14 1460 3629029
BC062563.1 - 14 1553 5140496

View Full Product Details