Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304957.1 | NP_001291886.1 | 5 | 814 | |
NM_001304958.1 | NP_001291887.1 | 6 | 921 | ||
NM_001304959.1 | NP_001291888.1 | 5 | 563 | ||
NM_001304960.1 | NP_001291889.1 | 7 | 732 | ||
NM_001304961.1 | NP_001291890.1 | 4 | 731 | ||
NM_025140.2 | NP_079416.1 | 5 | 824 | ||
XM_005253624.2 | XP_005253681.1 | 5 | 568 | ||
XM_017019984.1 | XP_016875473.1 | 5 | 1382 | ||
GenBank mRNA | AB015292.1 | - | 4 | 344 | |
AK026124.1 | - | 5 | 455 | ||
AK054680.1 | - | 3 | 2091 | ||
AK124358.1 | - | 1 | 1913 | ||
AK125866.1 | - | 3 | 2342 | ||
AK222661.1 | - | 5 | 412 | ||
AW953473.1 | - | 8 | 703 | ||
BC017186.2 | - | 5 | 447 | 3051346 | |
BX394686.2 | - | 4 | 480 | ||
DA370549.1 | - | 6 | 519 |