Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000242.2 | NP_000233.1 | 4 | 1837 | |
XM_006717861.3 | XP_006717924.1 | 5 | 1905 | ||
XM_011539816.2 | XP_011538118.1 | 5 | 1889 | ||
GenBank mRNA | X15422.1 | - | 4 | 1837 |