Gene Transcripts
- Gene Symbol
- PLAA
- Entrez Gene ID
- 9373
- Gene Name
- phospholipase A2 activating protein
- Gene Aliases
- DOA1, PLA2P, PLAP
- Chromosome Location
- Chr. 9: 26903370 - 26947473 on Build GRCh38
- UniGene ID
- Hs.27182
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031689.2 |
NP_001026859.1
|
2, 3 |
761 |
|
NM_001321546.1 |
NP_001308475.1
|
2, 3 |
764 |
|
XM_011518072.2 |
XP_011516374.1
|
2, 3 |
750 |
|
XM_017015319.1 |
XP_016870808.1
|
2, 3 |
750 |
|
GenBank mRNA |
AF083395.1 |
- |
1, 2 |
165 |
|
AF145020.1 |
- |
2, 3 |
560 |
|
AJ238243.1 |
- |
1, 2 |
193 |
|
AK001642.1 |
- |
2, 3 |
464 |
|
AK002143.1 |
- |
2, 3 |
524 |
|
AK022761.1 |
- |
2, 3 |
439 |
|
AK223544.1 |
- |
1, 2 |
196 |
|
AL133608.1 |
- |
2, 3 |
754 |
|
BC032551.1 |
- |
2, 3 |
443 |
5555038 |
View Full Product Details