Gene Transcripts
- Gene Symbol
- TRIP12
- Entrez Gene ID
- 9320
- Gene Name
- thyroid hormone receptor interactor 12
- Gene Aliases
- TRIP-12, ULF
- Chromosome Location
- Chr. 2: 229763837 - 229923234 on Build GRCh38
- UniGene ID
- Hs.572642
- Species
- Homo sapiens
View Full Product Details