Gene Transcripts

Gene Symbol
CLPTM1
Entrez Gene ID
1209
Gene Name
CLPTM1, transmembrane protein
Gene Aliases
Chromosome Location
Chr. 19: 44954585 - 44993346 on Build GRCh38
UniGene ID
Hs.444441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282175.1 NP_001269104.1 14 1817
NM_001282176.1 NP_001269105.1 14 1841
NM_001294.3 NP_001285.1 14 1968
GenBank mRNA AF037339.1 - 14 1811
AK027698.1 - 2 264
AK074935.1 - 14 1812
AK075262.1 - 14 1811
AK223553.1 - 14 1812
AK293314.1 - 14 1833
AK304480.1 - 14 1800
AK316094.1 - 14 1837
BC004865.2 - 14 1847 3840991
BC012359.1 - 14 1801 4685026
BT007262.1 - 14 1796
KU177986.1 - 14 1796
KU177987.1 - 14 1796

View Full Product Details