Gene Transcripts

Gene Symbol
EMID1
Entrez Gene ID
129080
Gene Name
EMI domain containing 1
Gene Aliases
EMI5, EMU1
Chromosome Location
Chr. 22: 29205912 - 29259597 on Build GRCh38
UniGene ID
Hs.289106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267895.1 NP_001254824.1 14 1298
NM_133455.3 NP_597712.2 14 1304
XM_011529869.2 XP_011528171.1 15 1309
XM_011529873.2 XP_011528175.1 15 1310
GenBank mRNA AJ416090.1 - 14 1171
AK097163.1 - 1 479
BC046358.1 - 14 1252 5770546
CR456407.1 - 14 1187

View Full Product Details