Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008924.2 | NP_032950.1 | 2, 3 | 605 | |
GenBank mRNA | AF533977.1 | - | 2, 3 | 605 | |
AK132728.1 | - | 2, 3 | 544 | ||
BC075623.1 | - | 2, 3 | 576 | 6854057 | |
BC080276.1 | - | 2, 3 | 599 | 30354403 | |
J02935.1 | - | 2, 3 | 288 |