Gene Transcripts

Gene Symbol
L3mbtl3
Entrez Gene ID
237339
Gene Name
l(3)mbt-like 3 (Drosophila)
Gene Aliases
AI481284, MBT-1
Chromosome Location
Chr. 10: 26274470 - 26375989 on Build GRCm38
UniGene ID
Mm.338439
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172787.2 NP_766375.1 7, 8 766
GenBank mRNA AK031398.1 - 7, 8 752
AK045667.1 - 7, 8 767
AK049597.1 - 7, 8 801
AK129443.1 - 3, 4 311
BC082309.1 - 6, 7 636 30356672
BC085192.1 - 6, 7 871 30694487

View Full Product Details