Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176979.5 | NP_795953.2 | 4, 5 | 421 | |
GenBank mRNA | AK084654.1 | - | 4, 5 | 524 | |
AK085031.1 | - | 4, 5 | 560 | ||
AK129104.1 | - | 4, 5 | 533 | ||
BC049797.1 | - | 4, 5 | 421 | 6510112 |