Gene Transcripts

Gene Symbol
Itih1
Entrez Gene ID
16424
Gene Name
inter-alpha trypsin inhibitor, heavy chain 1
Gene Aliases
ITI-HC1, Intin1, Itih-1
Chromosome Location
Chr. 14: 30929180 - 30943321 on Build GRCm38
UniGene ID
Mm.3227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008406.3 NP_032432.2 5 510
GenBank mRNA AK149500.1 - 5 511
BC013465.1 - 5 494
BC028814.1 - 5 510 5099284
X70391.1 - 5 490

View Full Product Details