Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010894.2 | NP_035024.1 | 2 | 2229 | |
GenBank mRNA | AK005073.1 | - | 2 | 2230 | |
AK018781.1 | - | 2 | 2229 | ||
AK028055.1 | - | 2 | 2234 | ||
BC094611.1 | - | 3 | 1811 | 30361598 |