Gene Transcripts

Gene Symbol
Pax8
Entrez Gene ID
18510
Gene Name
paired box 8
Gene Aliases
Pax-8
Chromosome Location
Chr. 2: 24420551 - 24480092 on Build GRCm38
UniGene ID
Mm.2533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011040.4 NP_035170.1 6 755
GenBank mRNA AK050717.1 - 6 731
AK052610.1 - 6 731
BC020526.1 - 6 722 4239835
X57487.1 - 6 712

View Full Product Details