Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011045.2 | NP_035175.1 | 6 | 930 | |
GenBank mRNA | AK012730.1 | - | 6 | 934 | |
AK012932.1 | - | 6 | 930 | ||
AK088256.1 | - | 6 | 931 | ||
AK151238.1 | - | 6 | 931 | ||
AK151411.1 | - | 6 | 963 | ||
AK165217.1 | - | 6 | 931 | ||
AK168233.1 | - | 6 | 931 | ||
AK168596.1 | - | 6 | 934 | ||
BC005778.1 | - | 6 | 915 | 3602797 | |
BC010343.1 | - | 6 | 914 | 3602977 | |
X53068.1 | - | 6 | 929 |