Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159964.1 | NP_001153436.1 | 3 | 487 | |
NM_007943.3 | NP_031969.1 | 15 | 1436 | ||
GenBank mRNA | AK028361.1 | - | 635 | ||
AK083176.1 | - | 3 | 471 | ||
AY190610.1 | - | 15 | 1409 | ||
BC048783.1 | - | 15 | 1410 | 6466448 | |
CF533628.1 | - | 3 | 487 | ||
L21768.1 | - | 15 | 1409 |