Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162494.1 | NP_001155966.1 | 2 | 406 | |
NM_008056.3 | NP_032082.3 | 2 | 410 | ||
GenBank mRNA | AK088399.1 | - | 2 | 275 | |
BB613669.1 | - | 2 | 406 | ||
BC026150.1 | - | 2 | 399 | 3983985 | |
CD348842.1 | - | 2 | 376 | ||
U43319.1 | - | 1 | 236 |