Gene Transcripts

Gene Symbol
Gabrb1
Entrez Gene ID
14400
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit beta 1
Gene Aliases
AW061132, B230208N19Rik, Gabrb-1
Chromosome Location
Chr. 5: 71658056 - 72149572 on Build GRCm38
UniGene ID
Mm.38567
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008069.4 NP_032095.1 7 702
GenBank mRNA BC130258.1 - 7 706
BC157920.1 - 7 768 9007350

View Full Product Details