Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010511.2 | NP_034641.1 | 6 | 996 | |
GenBank mRNA | AF128216.1 | - | 6 | 947 | |
AF128217.1 | - | 6 | 947 | ||
AK087525.1 | - | 6 | 985 | ||
AK150503.1 | - | 6 | 945 | ||
AK151079.1 | - | 6 | 950 | ||
AK152085.1 | - | 6 | 945 | ||
AK152384.1 | - | 6 | 945 | ||
AK153084.1 | - | 6 | 945 | ||
AK153347.1 | - | 6 | 948 | ||
AK159359.1 | - | 6 | 950 | ||
AK159660.1 | - | 6 | 890 | ||
AK159839.1 | - | 6 | 890 | ||
AK169593.1 | - | 6 | 965 | ||
AY157836.1 | - | 6 | 947 | ||
AY157837.1 | - | 6 | 947 | ||
BC132599.1 | - | 6 | 897 | 40130876 | |
BC138617.1 | - | 6 | 897 | 8861637 | |
J05265.1 | - | 6 | 973 | ||
M25764.1 | - | 6 | 898 | ||
M26711.1 | - | 6 | 973 | ||
M28233.1 | - | 6 | 969 | ||
M28995.1 | - | 6 | 874 |