Gene Transcripts

Gene Symbol
Cdca3
Entrez Gene ID
14793
Gene Name
cell division cycle associated 3
Gene Aliases
2410005A12Rik, C8, Grcc8, Tome-1
Chromosome Location
Chr. 6: 124829595 - 124833701 on Build GRCm38
UniGene ID
Mm.285723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013538.5 NP_038566.1 6 1180
GenBank mRNA AK010409.1 - 6 1121
BC002006.1 - 6 1134 3592108
BC027172.1 - 5 1758

View Full Product Details