Gene Transcripts

Gene Symbol
Acad9
Entrez Gene ID
229211
Gene Name
acyl-Coenzyme A dehydrogenase family, member 9
Gene Aliases
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002
Chromosome Location
Chr. 3: 36065984 - 36092857 on Build GRCm38
UniGene ID
Mm.260997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC004737.1 - 1 150
BC031137.1 - 7 840 4971848
BC032213.1 - 7 845 5323723
BC033277.1 - 7 825 4241041

View Full Product Details